4-(Hydroxymethyl)phenylboronic acid pinacol ester

Supplier: Thermo Scientific Chemicals

Synonyms: 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl alcohol, 2-[4-(Hydroxymethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-(Hydroxymethyl)benzeneboronic acid pinacol ester, (4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanol, 4-(Hydroxymethyl)phenylboronic acid Pinacol ester

H51123-03 H51123-14 H51123-06
AAH51123-03EA 50.63 USD
AAH51123-03 AAH51123-14 AAH51123-06
4-(Hydroxymethyl)phenylboronic acid pinacol ester
4-(Hydroxymethyl)phenylboronic acid pinacol ester

CAS No.: 302348-51-2 Molecular Formula: C13H19BO3 Formula Weight: 234.10 MDL No.: MFCD09837617

Formula: C₁₃H₁₉BO₃
MW: 234.10 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09837617
CAS Number: 302348-51-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR