2-(Trifluoromethyl)acrylic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-(Trifluoromethyl)propenoic acid

B22443-03 B22443-14 B22443-06
AAB22443-03EA 64.6 USD
AAB22443-03 AAB22443-14 AAB22443-06
2-(Trifluoromethyl)acrylic acid 98%
2-(Trifluoromethyl)acrylic acid

Purity: 98% CAS No.: 381-98-6 Molecular Formula: C4H3F3O2 Formula Weight: 140.06 Flash Point: 82°(180°F) Melting Point: 49-51° UN No.: 3261 MDL No.: MFCD00042424

Formula: C₄H₃F₃O₂
MW: 140.06 g/mol
Melting Pt: 49…51 °C
Flash Pt: 82 °C(180 °F)
Storage Temperature: Ambient
MDL Number: MFCD00042424
CAS Number: 381-98-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR