(S)-(-)-N-Benzyl-α-methylbenzylamine ≥99%, ee 99%, ChiPros®

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: S-(-)-N-Benzyl-1-phenylethylamine, (S)-(-)-N-Benzyl-1-phenylethylamine

L19299-14 L19299-06
AAAL19299-14EA 141 USD
AAAL19299-14 AAAL19299-06
(S)-(-)-N-Benzyl-α-methylbenzylamine ≥99%, ee 99%, ChiPros®
(S)-(-)-N-Benzyl-α-methylbenzylamine

Purity: 99% CAS No.: 17480-69-2 Molecular Formula: C15H17N Formula Weight: 211.31 Flash Point: 110°(230°F) Refractive Index: 1.5640 Boiling Point: 171°/15mm Beilstein Reference: 3650264 Optical Rotation: -39° (neat) MDL No.: MFCD00066325

Formula: C₁₅H₁₇N
MW: 211.31 g/mol
Boiling Pt: 171 °C/15 mm
Density: 1.010
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00066325
CAS Number: 17480-69-2
EINECS: 605-736-1
UN: 2735
ADR: 8 III

Order Now

Specification Test Results

Optical rotation -39° (neat)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR