(R)-(+)-α-Methylbenzylamine ≥99%, ee 99%, ChiPros®

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (R)-(+)-α-Phenylethylamine, (R)-(+)-1-Phenylethylamine, D(+)-α-Methylbenzylamine

L19117-22 L19117-14 L19117-06
AAAL19117-22EA 179 USD
AAAL19117-22 AAAL19117-14 AAAL19117-06
(R)-(+)-α-Methylbenzylamine ≥99%, ee 99%, ChiPros®
(R)-(+)-α-Methylbenzylamine

Purity: 99+% CAS No.: 3886-69-9 Molecular Formula: C8H11N Formula Weight: 121.18 Flash Point: 71°(159°F) Refractive Index: 1.5260 Odor: Amine-like Melting Point: -10° Boiling Point: 187° Beilstein Reference: 2410916 Merck Reference: 14,6026 Optical Rotation: +30° (c=10 in ethanol) UN No.: 2735 MDL No.: MFCD00064405

Formula: C₈H₁₁N
MW: 121.18 g/mol
Boiling Pt: 187 °C
Melting Pt: –10 °C
Density: 0.948
Flash Pt: 71 °C (159 °F)
Storage Temperature: Ambient
MDL Number: MFCD00064405
CAS Number: 3886-69-9
EINECS: 223-423-4
UN: 2922
ADR: 8 III
Merck Index: 14,6026

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR