Diallyl maleate 95%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Diallyl Maleate

L03867-30 L03867-0B
AAAL03867-18EA 46.86 USD
AAAL03867-18 AAAL03867-30 AAAL03867-0B
Diallyl maleate 95%
Diallyl maleate

Purity: 95% CAS No.: 999-21-3 Molecular Formula: C10H12O4 Formula Weight: 196.21 Flash Point: 130°(266°F) Refractive Index: 1.4670 Melting Point: -47° Boiling Point: 109-110°/3mm Beilstein Reference: 1725954 UN No.: 2810 MDL No.: MFCD00010763

Formula: C₁₀H₁₂O₄
MW: 196.21 g/mol
Boiling Pt: 109…110 °C (3 mmHg)
Melting Pt: –47 °C
Density: 1.072
Flash Pt: 130 °C (266 °F)
Storage Temperature: Ambient
MDL Number: MFCD00010763
CAS Number: 999-21-3
EINECS: 213-658-0
UN: 2810
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR