Diethyl allylmalonate 96%

Supplier: Thermo Scientific Chemicals
Warning

L02286-22 L02286-14
AAAL02286-22EA 117.21 USD
AAAL02286-22 AAAL02286-14
Diethyl allylmalonate 96%
Diethyl allylmalonate

Purity: 96% CAS No.: 2049-80-1 Molecular Formula: C10H16O4 Formula Weight: 200.23 Flash Point: 92°(197°F) Refractive Index: 1.4310 Boiling Point: 222-223° Beilstein Reference: 1709466 MDL No.: MFCD00009155

Formula: C₁₀H₁₆O₄
MW: 200.23 g/mol
Boiling Pt: 222…223 °C
Density: 1.016
Flash Pt: 92 °C (197 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009155
CAS Number: 2049-80-1
EINECS: 218-072-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR