1H,1H,9H-Hexadecafluoro-1-nonanol 97%

Supplier: Thermo Scientific Chemicals

Synonyms: 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9-Hexadecafluoro-1-nonanol, 1H,1H,9H-Perfluoro-1-nonanol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9-Hexadecafluor-1-nonanol, 1H,1h,9h-hexadecafluorononanol

B21407-22
AAB21407-06EA 25.45 USD
AAB21407-06 AAB21407-14 AAAB21407-22
1H,1H,9H-Hexadecafluoro-1-nonanol 97%
1H,1H,9H-Hexadecafluoro-1-nonanol

Purity: 97% CAS No.: 376-18-1 Molecular Formula: H(CF2)8CH2OH Formula Weight: 432.11 Melting Point: 55-60° Boiling Point: 157-158°/200mm MDL No.: MFCD00039629

Formula: C₉H₄F₁₆O
MW: 432.1 g/mol
Boiling Pt: 157…158 °C/200 mm
Melting Pt: 55…60 °C
Storage Temperature: Ambient
MDL Number: MFCD00039629
CAS Number: 376-18-1
EINECS: 206-806-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR