Tetraethylammonium bromide 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Tetrylammonium bromide

A13835-0B A13835-0I A13835-30
AAAA13835-0BEA 248.5 USD
AAAA13835-0B AAAA13835-0I AAAA13835-30
Tetraethylammonium bromide 98%
Tetraethylammonium bromide

Phase transfer catalyst


Purity: 98% CAS No.: 71-91-0 Molecular Formula: (CH3CH2)4NBr Formula Weight: 210.16 Odor: Mild Melting Point: ca 285° dec. Beilstein Reference: 3563430 Merck Reference: 14,9199 MDL No.: MFCD00011825

Formula: C₈H₂₀BrN
MW: 210.16 g/mol
Melting Pt: ∼ 285 °C (dec.)
Density: 1.397
Storage Temperature: Ambient
MDL Number: MFCD00011825
CAS Number: 71-91-0
EINECS: 200-769-4
Merck Index: 14,9199

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR