(-)-Quinine, anhydrous 99% (total base) may cont. up to 5% dihydroquinine

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (R)-[(2S,4S,5R)-1-Aza-5-vinylbicyclo[2.2.2]oct-2-yl](6-methoxyquinolin-4-yl)methanol, Quinine, (R)-(6-methoxyquinolin-4-yl)((1S,2S,4S,5R)-5-vinylquinuclidin-2-yl)methanol

A10459-09 A10459-30 A10459-18
AAAA10459-09EA 67.7 USD
AAAA10459-09 AAAA10459-30 AAAA10459-18
(-)-Quinine, anhydrous 99% (total base) may cont. up to 5% dihydroquinine
(-)-Quinine

A potassium channel blocker


Purity: 99% CAS No.: 130-95-0 Molecular Formula: C20H24N2O2 Formula Weight: 324.42 Melting Point: ca 177° dec. Beilstein Reference: 91867 Merck Reference: 14,8061 Optical Rotation: -166° (c=1 in ethanol) MDL No.: MFCD00198096

Formula: C₂₀H₂₄N₂O₂
MW: 324.42 g/mol
Melting Pt: 176 (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00198096
CAS Number: 130-95-0
EINECS: 205-003-2
Merck Index: 14,8061

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR