Ethyl hydrogen malonate ≥90%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Mono-Ethyl malonate, 3-Ethoxy-3-oxopropanoic acid

044692-14 044692-22 044692-06 044692-36
AAA44692-14EA 120 USD
AAA44692-14 AAA44692-22 AAA44692-06 AAA44692-36
Ethyl hydrogen malonate ≥90%
Ethyl hydrogen malonate

Liquid


Purity: 90+% CAS No.: 1071-46-1 Molecular Formula: C5H8O4 Formula Weight: 132.12 Flash Point: 110°(230°F) Physical Form: Liquid Density: 1.119 Refractive Index: 1.4295 Appearance: Pale yellow Boiling Point: 106°/3mm Beilstein Reference: 1758845 MDL No.: MFCD00020490

Formula: C₅H₈O₄
MW: 132.115 g/mol
Boiling Pt: 106 °C (3 mmHg)
Density: 1.119
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00020490
CAS Number: 1071-46-1
EINECS: 213-992-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR