1-Ethylpiperidine 99%

Supplier: Thermo Scientific Chemicals
Danger

A16294-36
AAA16294-36EA 116 USD
AAA16294-36
1-Ethylpiperidine 99%
1-Ethylpiperidine

Purity: 99% CAS No.: 766-09-6 Molecular Formula: C7H15N Formula Weight: 113.20 Flash Point: 18°(64°F) Refractive Index: 1.4440 Melting Point: -20° Boiling Point: 129-131° Beilstein Reference: 102643 UN No.: 2386 MDL No.: MFCD00006507

Formula: C₇H₁₅N
MW: 113.2 g/mol
Boiling Pt: 131
Melting Pt: -57
Density: 0.825
Flash Pt: 18 °C (64 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006507
CAS Number: 766-09-6
EINECS: 212-161-6
UN: 2386
ADR: 3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR