Methyl crotonate all trans 98% (by GC), liquid

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Methyl trans-2-butenoate, 2-Butenoic acid methyl ester, trans-Methyl crotonate, trans-Crotonic acid methyl ester, Crotonic acid methyl ester, trans-2-Butenoic acid methyl ester

A15363-22 A15363-36
AAA15363-22EA 45.37 USD
AAA15363-22 AAA15363-36
Methyl crotonate all trans 98% (by GC), liquid
Methyl crotonate

Purity: 98% CAS No.: 623-43-8 Molecular Formula: C5H8O2 Formula Weight: 100.12 Flash Point: 4°(40°F) Refractive Index: 1.4245 Boiling Point: 118-120° Beilstein Reference: 1720292 Merck Reference: 14,2597 UN No.: 3272 MDL No.: MFCD00009287

Formula: C₅H₈O₂
MW: 100.12 g/mol
Boiling Pt: 118…120 °C
Density: 0.946
Flash Pt: 4 °C (40 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009287
CAS Number: 623-43-8
EINECS: 210-793-7
UN: 3272
ADR: 3 III
Merck Index: 14,2597

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR