2-Methylcyclohexanone 98%

Supplier: Thermo Scientific Chemicals
Warning

A14741-0F A14741-AE A14741-AP
AAA14741-0FEA 1942 USD
AAA14741-0F AAA14741-AE AAA14741-AP
2-Methylcyclohexanone 98%
2-Methylcyclohexanone

Purity: 98% CAS No.: 583-60-8 Molecular Formula: C7H12O Formula Weight: 112.17 Flash Point: 46°(114°F) Physical Form: Clear liquid Density: 0.925 Refractive Index: 1.4480 Appearance: Colorless Melting Point: -14° Boiling Point: 162-163° Beilstein Reference: 506751 UN No.: 2297 MDL No.: MFCD00001635

Formula: C₇H₁₂O
MW: 112.17 g/mol
Boiling Pt: 162…163 °C
Melting Pt: –14 °C
Density: 0.925
Flash Pt: 46 °C (114 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001635
CAS Number: 583-60-8
EINECS: 209-513-6
UN: 2297
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR