N,N'-Diallyl-L-tartardiamide 99%

Supplier: Thermo Scientific Chemicals
Warning

A12195-18 A12195-30
AAA12195-18EA 88.96 USD
AAA12195-18 AAA12195-30
N,N'-Diallyl-L-tartardiamide 99%
N,N'-Diallyl-L-tartardiamide

Applications: Cross-linking co-monomer for the polymerization of soluble polyacrylamide gels


Purity: 99% CAS No.: 58477-85-3 Molecular Formula: C10H16N2O4 Formula Weight: 228.25 Melting Point: 184-186° Beilstein Reference: 1712934

Formula: C₁₀H₁₆N₂O₄
MW: 228.25 g/mol
Melting Pt: 184…186 °C
Storage Temperature: Ambient
MDL Number: MFCD00008640
CAS Number: 58477-85-3
EINECS: 261-277-3

Order Now

Specification Test Results

Optical rotation +118° (c=3 in methanol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR