Isonicotinamidine hydrochloride 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Pyridine-4-carboxamidine hydrochloride, Isonicotinimidamide hydrochloride, 4-Carbamimidoylpyridine hydrochloride, 4-Amidinopyridine hydrochloride, Pyridine-4-carboximidamide hydrochloride, 4-Amidinopyridinium chloride

A11050-06 A11050-14
AAA11050-03EA 34.76 USD
AAA11050-03 AAA11050-06 AAA11050-14
Isonicotinamidine hydrochloride 98+%
Isonicotinamidine hydrochloride

Purity: 98+% CAS No.: 6345-27-3 Molecular Formula: C6H7N3.HCl Formula Weight: 157.60 Melting Point: ca 245° dec. Beilstein Reference: 3562678

Formula: C₆H₇N₃·ClH
MW: 157.601 g/mol
Melting Pt: ∼ 245 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00040385
CAS Number: 6345-27-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR