Dichloro bis(dicyclohexylphosphino)propane palladium(II) (17.3% Pt)

Supplier: Thermo Scientific Chemicals
Warning

046780-03
AA46780-03EA 342 USD
AA46780-03
Dichloro bis(dicyclohexylphosphino)propane palladium(II) (17.3% Pt)
Dichloro bis(dicyclohexylphosphino)propane palladium(II)

Dichloro[bis(dicyclohexylphosphino)propane]palladium(II), Pd 17.3%


CAS No.: 1041005-52-0 Molecular Formula: C27H50Cl2P2Pd Formula Weight: 613.97 Physical Form: Crystalline solid Appearance: White MDL No.: MFCD14155689

Formula: C₂₇H₅₀Cl₂P₂Pd
MW: 613.965 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14155689
CAS Number: 1041005-52-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR