Tantalum ≥99.9% (metals basis), pellets, Diameter 1 mm (0.04 in) x 2 mm (0.08 in)

Supplier: Thermo Scientific Chemicals

044367-KO
AA44367-KOEA 1126 USD
AA44367-KO
Tantalum ≥99.9% (metals basis), pellets, Diameter 1 mm (0.04 in) x 2 mm (0.08 in)
Tantalum

Purity: 99.9% CAS No.: 7440-25-7 Odor: Odorless MDL No.: MFCD00011252

Formula: Ta
MW: 180.95 g/mol
Boiling Pt: 5427
Melting Pt: 2996
Density: 16.69 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011252
CAS Number: 7440-25-7
EINECS: 231-135-5
Merck Index: 12,09223

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR