Zinc oxide ≥99.9% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Zinci oxidum, Zinc (II) oxide

044263-A1 044263-A7 044263-30
AA44263-A1EA 106 USD
AA44263-A1 AA44263-A7 AA44263-30
Zinc oxide ≥99.9% (metals basis)
Zinc oxide

MDL: MFCD00011300


Purity: 99.9% CAS No.: 1314-13-2 Molecular Formula: ZnO Formula Weight: 81.37 Physical Form: -200 Mesh Powder Density: 5.606 Melting Point: 1975° Merck Reference: 14,10147 UN No.: 3077 MDL No.: MFCD00011300

Formula: ZnO
MW: 81.39 g/mol
Melting Pt: 1975 °C
Density: 5.606
Storage Temperature: Ambient
MDL Number: MFCD00011300
CAS Number: 1314-13-2
EINECS: 215-222-5
UN: 3077
ADR: 9 III
REACH: 01-2119463881-32
Merck Index: 14,10147

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR