Tetraethylammonium perchlorate ≥98% (dry weight) wetted with 10% water

Supplier: Thermo Scientific Chemicals
Danger

044062-09 044062-18
AA44062-09EA 86 USD
AA44062-09 AA44062-18
Tetraethylammonium perchlorate ≥98% (dry weight) wetted with 10% water
Tetraethylammonium perchlorate

Tetraethylammonium perchlorate is used as supporting electrolyte in polarographic measurements.


Purity: 98% CAS No.: 2567-83-1 Molecular Formula: (C2H5)4NClO4 Formula Weight: 229.71 Physical Form: Wet crystals Odor: Odorless Beilstein Reference: 3638093 UN No.: 1479 MDL No.: MFCD00041977

Formula: C₈H₂₀ClNO₄
MW: 229.7 g/mol
Melting Pt: >300 °C
Density: 1.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00041977
CAS Number: 2567-83-1
EINECS: 219-904-3
UN: 1479
ADR: 5.1 II

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR