Copper beryllium alloy (98.1:1.9 w%), alloy C17200, foil, Thickness 0.25 mm (0.01 in)

Supplier: Thermo Scientific Chemicals
Danger

041913-GU
AA41913-GUEA 400 USD
AA41913-GU
Copper beryllium alloy (98.1:1.9 w%), alloy C17200, foil, Thickness 0.25 mm (0.01 in)
Copper beryllium alloy (98.1:1.9 w%), alloy C17200

MDL: MFCD01320550


Molecular Formula: Cu:Be; 98.1:1.9 wt% Physical Form: ≈62.8g/150x200mm Odor: Odorless MDL No.: MFCD01320550

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR