o-Xylene-[D₁₀] (98% D) (Isotopic) for NMR spectroscopy

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,2-Dimethylbenzene-D10, o-Xylene-[²H₁₀]

036472-06
AA36472-06EA 271.95 USD
AA36472-06
o-Xylene-[D₁₀] (98% D) (Isotopic) for NMR spectroscopy
o-Xylene-[D₁₀]

MDL: MFCD00037698 Notes: NMR Solvent


Purity: 98+% CAS No.: 56004-61-6 Molecular Formula: C8D10 Formula Weight: 116.25 Physical Form: Liquid Density: 0.953 Appearance: Colorless Melting Point: -25° Boiling Point: 143-145° Merck Reference: 14,10081 UN No.: 1307 MDL No.: MFCD00037698

Formula: C₆D₄(CD₃)₂
MW: 116.09 g/mol
Boiling Pt: 142 °C (1013 hPa)
Melting Pt: -25 °C
Density: 0.953 g/cm³ (20 °C)
Flash Pt: 30 °C
Storage Temperature: Ambient
MDL Number: MFCD00037698
CAS Number: 56004-61-6
EINECS: 259-942-8
UN: 1307
ADR: 3,III
Merck Index: 14,10081

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR