Iron(III) sulfate hydrate, REacton®, Reagent Grade

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ferric Persulphate, Diiron trissulphate hydrate, Ferric sulphate hydrate

033316-30 033316-A1 033316-A7
AA33316-30EA 71.37 USD
AA33316-30 AA33316-A1 AA33316-A7
Iron(III) sulfate hydrate, REacton®, Reagent Grade
Iron(III) sulphate hydrate

MDL: MFCD00011007 Soluble in water


CAS No.: 15244-10-7 Molecular Formula: Fe2(SO4)3.xH2O Formula Weight: 399.87(anhy) Physical Form: Granular/Powder Density: 2.0-2.1 Sensitivity: Hygroscopic Appearance: Grayish white Solubility: Soluble in water MDL No.: MFCD00011007

Formula: Fe₂(SO₄)₃·H₂O
MW: 399.88 g/mol (anhy)
Density: 2.0…2.1
Storage Temperature: Ambient
MDL Number: MFCD00011007
CAS Number: 15244-10-7
EINECS: 233-072-9
Merck Index: 13,04065

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR