Ammonium cerium(IV) nitrate ≥98.5% ACS

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Ammonium ceric nitrate, Ammonium hexanitratocerate (IV), Ceric Ammonium Nitrate, Cerium (IV) ammonium nitrate, Diammonium hexanitratocerate

033254-36 033254-A3 033254-22
AA33254-36EA 335.03 USD
AA33254-36 AA33254-A3 AA33254-22
Ammonium cerium(IV) nitrate ≥98.5% ACS
Ammonium cerium(IV) nitrate

Analytical chemistry, as an oxidant for organic compounds, as a polymerization catalyst for olefins, and as a scavenger in the manufacture of azides


Purity: 98.5% min CAS No.: 16774-21-3 Molecular Formula: H8CeN8O18 Formula Weight: 548.23 Physical Form: Granular Appearance: Yellow to orange Odor: Pungent Solubility: Soluble in water and alcohol Merck Reference: 14,1992 UN No.: 1477 MDL No.: MFCD00151121


Analytical chemistry, as an oxidant for organic compounds, as a polymerization catalyst for olefins, and as a scavenger in the manufacture of azides

Formula: (NH₄)₂Ce(NO₃)₆
MW: 548.22 g/mol
Melting Pt: 107…108 °C
Density: 2.2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00151121
CAS Number: 16774-21-3
EINECS: 240-827-6
UN: 1477
ADR: 5.1 II
Merck Index: 14,1992

Order Now

Specification Test Results

Insoluble in dilute sulfuric acid 0.05 %
Cl 0.01%
PO4 0.02%
Fe 0.005%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR