Iron(II) bromide, dry_ultra 99,995 Metals basis

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Iron dibromide, Ferrous bromide, Iron(II) bromide

014541-03
AA14541-03EA 110.63 USD
AA14541-03
Iron(II) bromide, dry_ultra 99,995 Metals basis
Iron(II) bromide

MDL: MFCD00016081


Purity: 99.995% CAS No.: 7789-46-0 Molecular Formula: FeBr2 Formula Weight: 215.67 Physical Form: -10 Mesh Beads, Ampouled under argon Density: 4.636 Melting Point: 684° Merck Reference: 14,4042 UN No.: 3260 MDL No.: MFCD00016081

Formula: Br₂Fe
MW: 215.66 g/mol
Melting Pt: 684 °C
Density: 4.636
Storage Temperature: Ambient
MDL Number: MFCD00016081
CAS Number: 7789-46-0
EINECS: 232-168-8
UN: 3260
ADR: 8 III
Merck Index: 14,4042

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR