Nickel, Plasma Standard Solution, Specpure®, Ni 10000 µg/ml, Thermo Scientific

Supplier: Thermo Scientific Chemicals

014414-AD 014414-AP 014414-AE
AA14414-ADEA 78.97 USD
AA14414-AD AA14414-AP AA14414-AE
Nickel, Plasma Standard Solution, Specpure®, Ni 10000 µg/ml, Thermo Scientific
Standards Cationic Standards Nickel Standards

Molecular Formula: Ni in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011137

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR