Sodium, Plasma Standard Solution, Specpure®, Na 10,000 µg/ml, Thermo Scientific

Supplier: Thermo Scientific Chemicals

014400-AP 014400-AD 014400-AE
AA14400-APEA 285 USD
AA14400-AP AA14400-AD AA14400-AE
Sodium, Plasma Standard Solution, Specpure®, Na 10,000 µg/ml, Thermo Scientific
Standards Cationic Standards Sodium Standards

Molecular Formula: Na2CO3 in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00085307

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR