Iodine ≥99.5%, crystalline

Supplier: Thermo Scientific Chemicals
Warning

014248-A1 014248-14 014248-30 014248-22 014248-28
AA14248-A1EA 540.24 USD
AA14248-A1 AA14248-14 AA14248-30 AA14248-22 AA14248-28
Iodine ≥99.5%, crystalline
Iodine

Iodine, crystalline, 99.5%


Purity: 99.5% CAS No.: 7553-56-2 Molecular Formula: I2 Formula Weight: 253.81 Physical Form: Solid Density: 4.930 Odor: Irritating Melting Point: 183-186° subl. UN No.: 3495 MDL No.: MFCD00011355

Formula: I₂
MW: 253.81 g/mol
Boiling Pt: 184.3
Melting Pt: 183…186 °C (subl.)
Density: 4.930
Storage Temperature: Ambient
MDL Number: MFCD00011355
CAS Number: 7553-56-2
EINECS: 231-442-4
UN: 3495
ADR: 8 III
Merck Index: 13,05036

Order Now

Specification Test Results

Assay 99.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR