Rubidium perchlorate, anhydrous ≥99.5% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

011628-18
AA11628-18EA 237 USD
AA11628-18
Rubidium perchlorate, anhydrous ≥99.5% (metals basis)
Rubidium perchlorate

MDL: MFCD00016294


Purity: 99.5% CAS No.: 13510-42-4 Molecular Formula: RbClO4 Formula Weight: 184.94 Physical Form: Crystalline Refractive Index: 1.4701 Sensitivity: Hygroscopic Odor: Odorless UN No.: 1481 MDL No.: MFCD00016294

Formula: ClO₄Rb
MW: 184.92
Melting Pt: 281
Density: 2.878
Storage Temperature: Ambient
MDL Number: MFCD00016294
CAS Number: 13510-42-4
EINECS: 236-840-1
UN: 1481
ADR: 5.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR