Potassium iodide ≥99.995% (metals basis)

Supplier: Thermo Scientific Chemicals

Synonyms: Dipotassium diiodide, Kali iodide

010978-14 010978-06 010978-22
AA10978-14EA 151 USD
AA10978-14 AA10978-06 AA10978-22
Potassium iodide ≥99.995% (metals basis)
Potassium iodide

MDL: MFCD00011405 Fieser: 5,327 6,227


Purity: 99.995% CAS No.: 7681-11-0 Molecular Formula: KI Formula Weight: 166.01 Physical Form: Crystalline Density: 3.12 Refractive Index: 1.677 Sensitivity: Hygroscopic Odor: Odorless Melting Point: 681° Boiling Point: 1330° Merck Reference: 14,7643 MDL No.: MFCD00011405

Formula: Kl
MW: 166 g/mol
Boiling Pt: 1330
Melting Pt: 686
Density: 3.12
Storage Temperature: Ambient
MDL Number: MFCD00011405
CAS Number: 7681-11-0
EINECS: 213-659-4
Merck Index: 14,7643

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR