Anti-CD44/HCAM Monoclonal Antibody [clone: 156-3C11]

Supplier: Diagnostic Biosystems

MOB256-05 MOB256 PDM172
95024-350EA 483.46 USD
95024-350 99990-490 10064-996
Anti-CD44/HCAM Monoclonal Antibody [clone: 156-3C11]
Antibodies
This antibody is specific to a cell surface glycoprotein of 80-95 kDa, known as CD44. CD44 is expressed on the surface of lymphocytes, monocytes and granulocytes. This antibody reacts with red cells pretreated with 2-aminoethlyl isothiouronium bromide (AET).

Type:
Antigen: CD44/HCAM
Clonality: Monoclonal
Clone: 156-3C11
Conjugation:
Epitope:
Host:
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR