VWR® Calibrated Electronic Thermometers with Waterproof Sensor
Supplier: VWR International
82020-064 82021-172 98000-208
82020-064EA 74.79 USD
82020-064 82021-172 98000-208
VWR® Calibrated Electronic Thermometers with Waterproof Sensor
Thermometers Digital Thermometers
Digital thermometers that effectively measure the temperature of wet or dry environments including freezers, refrigerators, water baths, aquariums, incubators and more.
  • Features available include minimum/maximum temperature readings, dual/triple display, and alarms
  • All models include accredited calibration documentation that meets the requirements of ISO 17025
  • Includes multi-language instructions (English, French, German, Italian, Portuguese, and Spanish)
  • Individually serialized, CE marked
Tested and calibrated in an exclusive triple accredited/More Product Information