Diaphragm Vacuum Pumps, Labconco®
Supplier: Labconco
5248200 5248201 7393000 7393001 7359020 7359030
97002-432EA 2125 USD
97002-432 97002-434 97109-870 97109-872 75796-634 75796-638
Diaphragm Vacuum Pumps, Labconco®
Pumps Vacuum Pumps
Low-maintenance, oil-free diaphragm vacuum pumps provide a reliable vacuum source for a variety of laboratory products including CentriVap Centrifugal Concentrators, FreeZone Freeze Dry Systems, RapidVap Vacuum Evaporation Systems, and Protector and Precise Controlled Atmosphere Glove Boxes.
These pumps require no oil, providing cleaner operation
VACUUBRAND Diaphragm Vacuum Pumps are sized for use with CentriVap Centrifugal Concentrators, RapidVap Vacuum Evaporation Systems and Vacuum Desiccators and have uses with many other pieces of laboratory equipment that require a vacuum source.More Product Information