Iodoacetamide 98, Proteomics grade

Supplier: VWR
Danger

Synonyms: 2-Iodoacetamide

M216-30G
97064-926CS 3906.74 USD
97064-926
Iodoacetamide 98, Proteomics grade
Iodoacetamide
Formula: ICH₂CONH₂
MW: 184.96 g/mol
Boiling Pt: 266
Melting Pt: 91.0 °C…93.0 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008028
CAS Number: 144-48-9
EINECS: 205-630-1
UN: 2928
ADR: 6.1 II

Order Now

Specification Test Results

Loss on Drying < 1%
Melting Point 90°C - 95°C
Protease none detected
Purity > 98 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR