Silver nitrate, Proteomics Grade

Supplier: VWR
Danger

Synonyms: Lunar caustic, Silver mononitrate, Nitric acid silver salt

M122-100G M122-500G
97064-872EA 775.92 USD
97064-872 97064-876
Silver nitrate, Proteomics Grade
Silver nitrate
Formula: AgNO₃
MW: 169.87 g/mol
Boiling Pt: 440 °C
Melting Pt: 212 °C
Density: 4.35 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003414
CAS Number: 7761-88-8
EINECS: 231-853-9
UN: 1493
ADR: 5.1 II
REACH: 01-2119513705-43
Merck Index: 12,08661
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f56d5ff35f4c0937041d9629e42aae69bf1987b3&vid=7ca52a55b378bddc27fb4a51ace3ae27a0451c75&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Chloride < 0.0005%
Clarity of Solution Pass
Copper <= 0.0002 %
Free Acid Test Pass
Iron < = 0.0002 %
Lead < = 0.001 %
Purity > = 99 %
Substances not Precipitated by HCl < 0.01 %
Sulfate <= 0.002 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR