D-(+)-Glucose 40% (w/v) in aqueous solution, Ultra Pure Grade for biochemistry

Supplier: VWR

Synonyms: Corn sugar, ᴅ-(+)-Glucose, Glucose, D-(+)-Dextrose, (2R,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal, Glucolin, Dextrose, D-Glucose

E701-100ML
97062-946EA 93.68 USD
97062-946
D-(+)-Glucose 40% (w/v) in aqueous solution, Ultra Pure Grade for biochemistry
D-(+)-Glucose
Formula: C₆H₁₂O₆
MW: 180.16 g/mol
Melting Pt: 146
MDL Number: MFCD00148912
CAS Number: 50-99-7
EINECS: 207-757-8
Merck Index: 14,04459
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=47ec28c7fd4803097405f8eaef3171b3a40d40c5&vid=62627c96e2ae9f2a0cae732d0ddf0a100247298e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear to slightly yellow liquid
Glucose 35 % - 40 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR