VWR® READY-PACK™ Ultra Pure Grade TRIS Buffered Saline (TBS)
Supplier: VWR
0788-2PK
97062-370EA 171.34 USD
97062-370
VWR® READY-PACK™ Ultra Pure Grade TRIS Buffered Saline (TBS)
Buffers Electrophoresis Buffers Washing Buffers
Pre-blended TBS buffer
Each Ready-Pack™ prepares 1 liter of a 20X solution of tris buffered saline when dissolved in water. A 1X solution contains 25mM Tris, 140mM Sodium Chloride, and 3.0mM Potassium Chloride.