HEPES sodium salt ≥99%, high purity

Supplier: VWR

Synonyms: Sodium 4-(2-hydroxyethyl)piperazin-1-ylethanesulphonate, Sodium 2-(4-(2-hydroxyethyl)piperazin-1-yl)ethanesulphonate, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulphonic acid) sodium salt, 4-(2-Hydroxyethyl)piperazine-1-ethanesulphonic acid sodium salt, HEPES sodium, HEPES Na

0485-500G 0485-100G 0485-1KG
97061-772EA 30.92 USD
97061-772 97061-774 97061-770 80761-250
HEPES sodium salt ≥99%, high purity
HEPES sodium salt

Biological buffers are useful for cell culture in vitro, enzyme assays, and some electrophoretic applications at physiological pH.

Formula: C₈H₁₇N₂NaO₄S
MW: 260.29 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00036463
CAS Number: 75277-39-3
EINECS: 278-169-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=116e9b4f3c17d013ff2b7c9848c219943a725b3c&vid=8c800de68c4434c3c9d77107ff6779ef516bd851&prid=web&clid=SAPDAM

Order Now

Specification Test Results

DNase None Detected
Heavy Metals (as Pb) <0.0001%
Loss on Drying ≤3%
pKa 7.45 - 7.65
Protease None Detected
Purity (Dry Basis) >99%
RNase None
Solubility (5%, Water) Pass


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR