L-Alanine, powder, high purity

Supplier: VWR

Synonyms: (S)-(+)-Alanine, H-Ala-OH, L(+)-Alanine, Alanine, L-α-Alanine, A, H-L-Ala-OH, α-Alanine

0106-100G
97061-024CS 675.35 USD
97061-024 97061-028
L-Alanine, powder, high purity
L-Alanine

Non-animal origin. White crystalline powder


Nonpolar. Aliphatic amino acid. Non-essential amino acid. Common component of culture media. Involved in regulation of a variety of metabolic functions.

Formula: C₃H₇NO₂
MW: 89.09 g/mol
Boiling Pt: 189 °C (1013 hPa)
Melting Pt: 314°C-316°C
Density: 1.4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064410
CAS Number: 56-41-7
EINECS: 200-273-8
Merck Index: 13,00203

Order Now

Specification Test Results

Arsenic < = 0.00015 %
Ash <= 0.15 %
Chloride <= 0.05 %
Heavy Metals <= 0.001 %
Sulfate < 0.03 %
Iron < 0.003 %
Loss on Drying < 0.2 %
pH (5%, Water) @25°C 5.5 - 7
Purity > 99 %
Specific Rotation 13.7 -15.1 °

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR