Atomax™ Hollow Cathode Lamps, PerkinElmer
Supplier: PerkinElmer
N2025300 N2025301 N2025305 N2025306 N2025308 N2025310 N2025317 N2025326 N2025330 N2025333 N2025338 N2025339 N2025365 N2025302 N2025304 N2025307 N2025309 N2025311 N2025313 N2025314 N2025315 N2025316 N2025318 N2025319 N2025320 N2025323 N2025324 N2025327 N2025329 N2025331 N2025332 N2025334 N2025341 N2025344 N2025345 N2025346 N2025348 N2025351 N2025352 N2025354 N2025355 N2025356 N2025360 N2025361 N2025363 N2025366
97050-210EA 969.88 USD
97050-210 97050-212 97050-216 97050-220 97050-222 97050-226 97050-230 97050-244 97050-254 97050-262 97050-266 97050-270 97050-276 97050-282 97050-286 97050-288 97050-294 97050-328 97050-340 97050-214 97050-218 97050-224 97050-228 97050-232 97050-234 97050-236 97050-238 97050-240 97050-242 97050-246 97050-248 97050-250 97050-252 97050-256 97050-258 97050-260 97050-264 97050-268 97050-272 97050-274 97050-278 97050-280 97050-284 97050-290 97050-292 97050-296 97050-298 97050-300 97050-302 97050-304 97050-306 97050-308 97050-310 97050-312 97050-314 97050-316 97050-318 97050-320 97050-322 97050-324 97050-326 97050-330 97050-332 97050-334 97050-336 97050-338 97050-342
Atomax™ Hollow Cathode Lamps, PerkinElmer
Lights Hollow Cathode Lamps
Single-element hollow cathode lamps are crafted to the highest standards.
  • Single-element design
  • High-quality construction
  • Near universal compatibility with atomic absorption instruments
Fit most commercially available atomic absorption instruments that require 37mm (11/2") diameter lamps.
Información para pedidos: Manufacturer's warranty for 5000 milliampere-hours per lamp; contact your Avantor representative for more information.