Test Sieves, Stainless Steel Wire, Verder Scientific
Supplier: Retsch
60169000038 60169000045 60169000053 60169000063 60169000075 60169000090 60169000106 60169000125 60169000150 60169000180 60169000212 60169000250 60169000300 60169000355 60169000425 60169000500 60169000600 60169000710 60169000850 60169001000 60169001180 60169001400 60169001700 60169002000 60169002360 60169002800 60169003350 60169004000 60169004750 60169005600 60169006300 60169006700 60169008000 60169009500 60169011200 60169012500 60169013200 60169016000 60169019000 60169022400 60169025000 60169026500 60169031500 60169037500 60169045000 60169050000 60169053000 60169063000 60169075000 60169090000 60169106000 60169125000
97034-572EA 1246.25 USD
97034-572 97034-574 97034-576 97034-578 97034-580 97034-582 97034-584 97034-586 97034-588 97034-590 97034-592 97034-594 97034-596 97034-598 97034-600 97034-602 97034-604 97034-606 97034-608 97034-610 97034-612 97034-614 97034-616 97034-618 97034-620 97034-622 97034-624 97034-626 97034-628 97034-630 97034-632 97034-634 97034-636 97034-638 97034-640 97034-642 97034-644 97034-646 97034-648 97034-650 97034-652 97034-654 97034-656 97034-658 97034-660 97034-662 97034-664 97034-666 97034-668 97034-670 97034-672 97034-674
Test Sieves, Stainless Steel Wire, Verder Scientific
Sieves Test Sieves
These test sieves consist of a solid stainless steel sieve frame of high stability for reliable sieving results. Sieves are mesh-specific and thus sieve fabric is precisely joined into the frame and tautened.
  • High degree of corrosion resistance and easy cleaning
  • Permanently tight sieve fabric
  • Extensive optical inspection
  • Clear and precise labeling of the sieves with full traceability based on individualised laser engraving
  • Maximum stability and optimum sealing when used in sieve stacks thanks to the o-ring which is placed in the recess designed for this
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable