InLab® Intelligent Sensor Management (ISM®) pH/Conductivity Electrodes, METTLER TOLEDO
Supplier: Mettler Toledo
51344163 51344055 30014096 51344110 51343156 51344102 51344211 51344072 30014092 30014093 30014094 51344104 51344114 51344116 51344172 51344155
97025-884EA 875 USD
97025-884 97025-874 89207-248 97025-888 97044-112 97025-870 97025-878 97025-880 89207-252 89207-254 89207-256 97025-872 97025-890 97025-892 97025-882 97025-876
InLab® Intelligent Sensor Management (ISM®) pH/Conductivity Electrodes, METTLER TOLEDO
Electrodes Multi-Parameter Electrodes
ISM® is a sophisticated safety concept that leaves nothing to chance.
  • Intelligent sensor management system (ISM®) internal reference
  • Calibration automatically transferred between compatible METTLER TOLEDO meters
  • Certificate can be sent to printer or PC
  • If the electrode comes with a cable, it will be listed in the order section below
The ISM® technology is compatible with following METTLER TOLEDO Meters: SevenExcellence™, SevenDirect™, SevenGo pro™, SevenGo Duo pro™, and SevenCompact™ (discontinued in June 2022).
The electrode certificate with theMore Product Information