Mobile Equipment Table, Labconco®
Supplier: Labconco
3819100 3819101 3819102
97025-418EA 2115 USD
97025-418 97025-420 97025-422
Mobile Equipment Table, Labconco®
Furniture Tables and Workbenches
Mobile equipment table may be used to transport heavy equipment and to provide auxiliary bench space.
  • 5" diameter non-marking polyurethane casters with toe locks have bearings for quiet operation
  • Supports loads up to 800 pounds
With height adjustments from 28.75" to 35.75", it may be raised to the same height as most laboratory countertops. Solid epoxy 1.25" thick × 30" deep work surface and epoxy-coated 1.75" square tubular steel frame.