HGA Pyrocoated Graphite Tubes with L'vov Groove, PerkinElmer
Supplier: PerkinElmer
B0121092 B0137111 B3001254
45603-036PK 1137.79 USD
45603-036 45603-042 97012-820
HGA Pyrocoated Graphite Tubes with L'vov Groove, PerkinElmer
Tubes Graphite Tubes
Standard pyrocoated graphite tubes are grooved to accept L'vov platforms and are produced in a quality controlled manner to ensure reliable temperature calibration in PerkinElmer graphite furnaces.
  • Pyrocoated to provide improved sensitivity and tube-to-tube reproducibility
  • Slotted to accept L'vov platforms (B3001256)
The tubes are compatible with PerkinElmer PinAAcle 900H, AAnalyst 700, HGA-900, HGA-800, HGA-700, HGA-600, HGA-300 AA instruments.