Sound Meter Type 1, Sper Scientific
Supplier: Sper Scientific
840015 840015C
97011-826EA 526.85 USD
97011-826 97011-828
Sound Meter Type 1, Sper Scientific
Sound Meters
Type 1 Sound Meter.
Features fast or slow time weightings, peak hold and AC output. A calibration adjustment allows the user to quickly calibrate the meter’s internal electronics. Can also be acoustically calibrated, in compliance with OSHA, using Acoustical Calibrator 850016 or equivalent.
Certificaciones: CE approved. Meets IEC61672 and ANSI S1.4 frequency weighting and time weighting specifications for a class 1 sound meter. N.I.S.T. traceable certificate of calibration (for certified model only).
Información accesorios: Simply place the calibrator on to the digital sound meter’sMore Product Information