VACUUBRAND® General-Purpose Oil-Free Diaphragm Vacuum Pumps, BrandTech
Supplier: Brandtech
20731000 20731003 20734000 20734003 20732000 20732003 20732200 20736000 20736003 20738000 20738003 20730003
97007-150EA 4825.89 USD
97007-150 97007-152 97007-154 97007-156 97007-158 97007-160 97007-162 97007-166 97007-168 97007-170 97007-172 89202-292 89511-182
VACUUBRAND® General-Purpose Oil-Free Diaphragm Vacuum Pumps, BrandTech
Pumps Vacuum Pumps
Designed specifically for non-corrosive and non-evaporative procedures, oil-free diaphragm vacuum pumps provide quiet and stable continuous-duty performance
  • For non-evaporative, non-corrosive applications
  • Integrated silencer(s)
  • High performance at working vacuum
  • 1-, 2- 3- and 4- stage designs
  • MD 4 and MZ 2D models feature KF16 flange inlets
  • All others have hose barbs
These aluminum-FKM diaphragm pumps utilize dry operation technology, eliminating the need to change or monitor oil. Diaphragms provide 10,000 to 15,000 hours of use before replacement,More Product Information