VWR® Models 1250, 1255 Glass Tube Flow Meters
Supplier: VWR International
97005-066 97005-070 97005-072 97005-076 97005-080 97005-082 97005-084 97005-086 97005-092 97005-094 97005-096 97005-108 97005-116 97005-138
97005-066EA 402.83 USD
97005-066 97005-070 97005-072 97005-076 97005-080 97005-082 97005-084 97005-086 97005-092 97005-094 97005-096 97005-108 97005-116 97005-138
VWR® Models 1250, 1255 Glass Tube Flow Meters
Flow Meters
Series 1250/55 are flowmeters for air or air/water and available in 65 or 150mm scale lengths.
  • Rotating lens provides a 180° view with magnification
  • Heavy-wall, precision bore
  • Borosilicate glass
  • Removeable without disconnnecting instrument
  • Threaded jack screw
EZ-CHANGE™ design allows quick interchangeability of tube assemblies. Flowmeters include ⅛" FNTP aluminum fittings and standard inlet valve (except 97005-138, which has ⅛" FNTP stainless steel fittings and precision inlet valve). Aluminum fittings include Buna seals.
Información para pedidos: More Product Information