Microplate Shakers, Boekel Scientific
Supplier: Boekel
270300 270340
97004-386EA 1413.05 USD
97004-386 97004-388
Microplate Shakers, Boekel Scientific
Shakers and Mixers Microplate Shakers
The Boekel Scientific Microplate Shaker features a low profile and small footprint and are available in a 2 Plate or 4 Plate Model
A simple screw attachment allows for quick and easy fitting of any standard depth 96 or 384 well plate while keeping your microplates securely in place. The LCD Display with microprocessor controls provide for easy viewing in the lab and includes a count down timer with auto shut off which emits an audible end of cycle alarm. This unit is also ideal for use in an incubator and cold rooms.
All Boekel Scientific products come with a 2-year warranty.