Compact Glove Boxes, Plas-Labs™
Supplier: Plas-Labs
830-CP-20 830-CP-30 830-CP-11 830-ABB 830-ABD 830-CP-10 830-CP-21 830-CP-31 830-ABC
97000-998EA 14426.52 USD
97000-998 97001-002 97000-996 10718-112 10718-114 97000-994 97001-000 97001-004 32940-008
Compact Glove Boxes, Plas-Labs™
Glove Boxes and Glove Bags
Designed specifically for use when weighing hazardous materials that must be contained, glove boxes provide cleanliness, safety, portability, and a draft-free atmosphere for the maximum effectiveness of your balance
Top section is removable for easy installation of equipment, such as analytical balances, top loaders, bench scales, and moisture analyzers. Basic features include four purging gas valves (two on the transfer chamber and two on the main chamber), a white leveling tray for easy transferring of liquids, white ambidextrous Hypalon® gloves for superior chemical and UV resistance,More Product Information