Space Drape, AIMS™
Supplier: Aims
SPDR
95059-642EA 20.23 USD
95059-642
Space Drape, AIMS™
First Aid Supplies Surgical Drapes
The space drape is designd to keep lab animals warm after the completion of procedures.
It features a soft flannel-like material on one side and a reflective coating on the other side.
Order Now
SPECIFICATIONS
Environmentally Preferable