Blocking Solution, 10% BSA, KPL Immunoassays
Supplier: KPL
5140-0006 5140-0008
95059-124EA 401.19 USD
95059-124 95059-126
Blocking Solution, 10% BSA, KPL Immunoassays
Buffers Electrophoresis Buffers Blocking Buffers
10% BSA Diluent/Blocking Solution minimizes non-specific binding and background
  • Readily dilutable
  • Convenient liquid format
  • Superior performance vs. non-fat dried milk
Provides excellent blocking on all membranes and plates. Suitable diluent for most immunossays.